Merck
  • Highly stable 7-N,N-dibutylamino-2-azaphenanthrene and 8-N,N-dibutylamino-2-azachrysene as a new class of second order NLO-active chromophores.
Highly stable 7-N,N-dibutylamino-2-azaphenanthrene and 8-N,N-dibutylamino-2-azachrysene as a new class of second order NLO-active chromophores.
Chemical communications (Cambridge, England) (2010-10-07)
Valentina Calabrese, Silvio Quici, Ester Rossi, Elena Cariati, Claudia Dragonetti, Dominique Roberto, Elisa Tordin, Filippo De Angelis, Simona Fantacci
ABSTRACT
7-N,N-Dibutylamino-2-azaphenanthrene (L(3)), 8-N,N-dibutylamino-2-azachrysene (L(4)) and related Ir(i) complexes or alkylated salts show high second-order NLO responses, as determined by the EFISH technique and DFT calculations. L(4) is appealing as building block for NLO active materials due to its unexpected large μβ(1.907)value and its very high thermal stability.
Product Description
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chrysene, analytical standard
Chrysene, BCR®, certified reference material