Merck
  • Impurity profiling of acetylspiramycin by liquid chromatography-ion trap mass spectrometry.
Impurity profiling of acetylspiramycin by liquid chromatography-ion trap mass spectrometry.
Journal of chromatography. A (2010-09-17)
Ming-juan Wang, Murali Pendela, Chang-qin Hu, Shao-hong Jin, Jos Hoogmartens, Ann Van Schepdael, Erwin Adams
ABSTRACT
Investigation of acetylspiramycin (ASPM) and its related substances was carried out using a reversed-phase liquid chromatography/tandem mass spectrometry method. The identification of impurities in the ASPM complex was performed with a quadrupole ion trap mass spectrometer, with an electrospray ionization (ESI) source in the positive ion mode which provides MS(n) capability. A total of 83 compounds were characterized in commercial samples, among which 31 impurities that had never been reported and 31 partially characterized impurities were deduced using the collision-induced dissociation (CID) spectra of major ASPM components as templates. Most of the major impurities arise from the starting materials and the synthesis process. This work provides very useful information for quality control of ASPM and evaluation of its synthesis process.
Product Description
Supelco
Spiramycin from Streptomyces sp., VETRANAL®, analytical standard, mixture of isomers
Spiramycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Spiramycin