Merck
  • Transkarbams as transdermal permeation enhancers: effects of ester position and ammonium carbamate formation.
Transkarbams as transdermal permeation enhancers: effects of ester position and ammonium carbamate formation.
Bioorganic & medicinal chemistry letters (2010-04-13)
Michal Novotný, Alexandr Hrabálek, Barbora Janůsová, Jakub Novotný, Katerina Vávrová
ABSTRACT
Transkarbam 12, an ammonium carbamate formed by the reaction of dodecyl 6-aminohexanoate with carbon dioxide, is a highly active, broad-spectrum, nontoxic, and nonirritant transdermal permeation enhancer. It probably acts by a dual mechanism: a part of its activity is associated with the carbamic acid salt and/or its decomposition in the acidic stratum corneum. The ammonium ester thereby released is an active enhancer species as well, and its activity highly depends on the position of the ester group.
Sigma-Aldrich
Ammonium carbamate, 99%
Supelco
Ammonium carbamate, for decomposition