Merck
  • Dose-response function for the soiling of silica-soda-lime glass due to dry deposition.
Dose-response function for the soiling of silica-soda-lime glass due to dry deposition.
The Science of the total environment (2009-11-11)
T Lombardo, A Ionescu, A Chabas, R-A Lefèvre, P Ausset, Y Candau
ABSTRACT
Several exposure campaigns of silica-soda-lime window glass have been performed in 30 European sites and 1 in Canada in order to understand, quantify and model the phenomenon of soiling. In this purpose samples were exposed sheltered from the rain. Parallel to exposure, several meteorological parameters and pollution concentrations have been monitored. This paper shows first results on the establishment of a dose-response function for glass soiling. Statistical analyses show that PM(10) is not the only parameter, but also SO(2) and NO(2) atmospheric concentrations seem to be responsible for the optical impairment of glass surfaces, expressed as haze.
Sigma-Aldrich
Soda lime, ACS reagent, granular, +100 mesh particle size
Sigma-Aldrich
Soda lime, p.a., with indicator, pellets
Sigma-Aldrich
Soda lime, for absorption of carbon dioxide