Merck
Patented small molecules against psoriasis.
Expert opinion on therapeutic patents (2009-07-02)
Alexander M Abdelnoor
ABSTRACT
It is hypothesized that psoriasis is an autoimmune disease. The most recent therapeutic approach that proved to be more effective than earlier methods of treatment is the use of mAb/fusion proteins. Efforts nowadays are focused on investigating the antipsoriatic affect of small molecules that can be administered orally, some of which are capable of entering cells, and being selective in targeting intracellular pathways. Preclinical patented small molecules that are recommended for the treatment of psoriasis are reviewed. Emphasis is placed on their mechanism of action. http://ep.espacenet.com/ , Pubmed, Scopus and Google websites were the main sources used for the patented small molecule search. A number of patents were poorly described and difficulties were faced in trying to figure out the patentee(s) explanation. Moreover, most patents were recommended for the treatment of a number of autoimmune diseases and cancer, and not only for psoriasis. Small molecules that inhibit the activation of T lymphocytes, leukocyte trafficking, leukotriene activity/production and angiogenesis, and promote apoptosis have been patented. Small molecules that have been patented for the treatment of other autoimmune diseases and could be used for treating psoriasis are described. Moreover, other possible mechanistic approaches using small molecules are discussed.
Sigma-Aldrich
Amygdalin, ≥99% (HPLC), from apricot kernels
Sigma-Aldrich
Amygdalin, BioXtra, ≥97.0% (HPLC)