Merck
  • Tricistronic overexpression of cytochrome P450cam, putidaredoxin, and putidaredoxin reductase provides a useful cell-based catalytic system.
Tricistronic overexpression of cytochrome P450cam, putidaredoxin, and putidaredoxin reductase provides a useful cell-based catalytic system.
Biotechnology letters (2009-05-22)
Donghak Kim, Paul R Ortiz de Montellano
ABSTRACT
The catalytic turnover of cytochrome P450( cam ) from Pseudomonas putida requires two auxiliary reduction partners, putidaredoxin (Pd) and putidaredoxin reductase (PdR). We report the functional expression in Escherichia coli of tricistronic constructs consisting of P450( cam ) encoded by the first cistron and the auxiliary proteins, Pd and PdR by the second and the third. Transformed bacterial whole cells efficiently oxidized (1R)-(+)-camphor to 5-exo-hydroxycamphor and, interestingly, limonene to (-)-perillyl alcohol. These bioengineered E. coli cells possess a heterologous self-sufficient P450 catalytic system that may have advantages in terms of low cost and high yield for the production of fine chemicals.
Sigma-Aldrich
(S)-(−)-Perillyl alcohol, 96%
Sigma-Aldrich
(S)-(−)-Perillyl alcohol, ≥95%, FG
Supelco
(−)-Perillyl alcohol, analytical standard