Merck
  • Structure and dissolution investigation of calcium-bismuth-borate glasses and vitroceramics containing silver.
Structure and dissolution investigation of calcium-bismuth-borate glasses and vitroceramics containing silver.
Journal of materials science. Materials in medicine (2007-03-06)
V Simon, M Spinu, R Stefan
ABSTRACT
Quaternary Ag(2)O-CaO-Bi(2)O(3)-B(2)O(3) glasses and glassceramics are investigated with regard to release behaviour and local structure. The dissolution behaviour in water and physiological serum shows that the cations are released rapidly or gradually and points out a multi-step process, generally characterised by higher rates in water than in physiological serum. The structural effect of silver addition to bismuth-borate glasses is observed from infrared spectroscopic data. The antibacterial activity of the investigated samples was tested on six bacterial media.
Sigma-Aldrich
Boric anhydride, 99.999% trace metals basis
Supelco
Boric anhydride, 99.98% trace metals basis
Sigma-Aldrich
Boric anhydride, granulated, ≥98.0% (T)
Sigma-Aldrich
Boric anhydride, puriss. p.a., ≥98% (T)