Merck
  • Analytical method for the determination of metolachlor, acetochlor, alachlor, dimethenamid, and their corresponding ethanesulfonic and oxanillic acid degradates in water using SPE and LC/ESI-MS/MS.
Analytical method for the determination of metolachlor, acetochlor, alachlor, dimethenamid, and their corresponding ethanesulfonic and oxanillic acid degradates in water using SPE and LC/ESI-MS/MS.
Analytical chemistry (2002-08-15)
Robert A Yokley, Louis C Mayer, Sung-Ben Huang, John D Vargo
ABSTRACT
A Good Laboratory Practices (GLP) validated, multiresidue analytical method is presented for the determination of the chloroacetanilide herbicides metolachlor, acetochlor, and alachlor, the chloroacetamide herbicide dimethenamid, and their respective ethanesulfonic (ESA) and oxanillic (OA) acid degradates in ground and surface water. A 50-mL water sample is subjected to purification using a C-18 SPE column. The four parent components and their eight ESA and OA degradates are isolated using 80/20 methanol/water (v/v) for elution. The eluate is reduced to < 1.0 mL and reconstituted in 10/90 acetonitrile/water (v/v) to the desired final fraction volume. Final analysis is accomplished using liquid chromatography/electrospray ionization-mass spectrometry/mass spectrometry in the + (parent compounds) and - (ESA and OA degradates) ion modes by monitoring appropriate precursor/product ion pairs for each of the 12 analytes. The method limit of quantification is 0.10 ppb and the limit of detection is 0.125 ng injected for each analyte. Average procedural recovery data range from 95 to 105% for fortification levels of 0.10-100 ppb. The method validation study was performed following GLP guidelines.
Product Description
Supelco
Metolachlor OA, PESTANAL®, analytical standard
Supelco
Metolachlor ESA sodium salt, PESTANAL®, analytical standard
Sigma-Aldrich
Ethanesulfonic acid, 95%
Supelco
Dimethenamid, PESTANAL®, analytical standard