Merck
  • Structure-activity relationships of 8-styrylxanthines as A2-selective adenosine antagonists.
Structure-activity relationships of 8-styrylxanthines as A2-selective adenosine antagonists.
Journal of medicinal chemistry (1993-05-14)
K A Jacobson, C Gallo-Rodriguez, N Melman, B Fischer, M Maillard, A van Bergen, P J van Galen, Y Karton
ABSTRACT
A series of substituted 8-styryl derivatives of 1,3,7-alkylxanthines was synthesized as potential A2-selective adenosine receptor antagonists, and the potency at rat brain A1- and A2-receptors was studied in radioligand binding experiments. At the xanthine 7-position, only small hydrophobic substituents were tolerated in receptor binding. 7-Methyl analogues were roughly 1 order of magnitude more selective for A2 versus A1 receptors than the corresponding 7-H analogues. 1,3-Dimethylxanthine derivatives tended to be more selective for A2-receptors than the corresponding 1,3-diallyl, diethyl, or dipropyl derivatives. Substitutions of the phenyl ring at the 3-(monosubstituted) and 3,5-(disubstituted) positions were favored. 1,3, 7-Trimethyl-8-(3-chlorostyryl)xanthine was a moderately potent (Ki vs [3H]CGS 21680 was 54 nM) and highly A2-selective (520-fold) adenosine antagonist. 1,3,7-Trimethyl-8-[(3-carboxy-1-oxopropyl)amino] styryl]xanthine was highly A2-selective (250-fold) and of enhanced water solubility (max 19 mM). 1,3-Dipropyl-7-methyl-8-(3,5-dimethoxystyryl) xanthine was a potent (Ki = 24 nM) and very A2-selective (110-fold) adenosine antagonist.
Sigma-Aldrich
8-(3-Chlorostyryl)caffeine, ≥98% (HPLC), solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
5 g
Please contact Customer Service for Availability
€42.00
25 g
Please contact Customer Service for Availability
€189.00
100 g
Please contact Customer Service for Availability
€439.00
To order products, please contact your local dealer.