Merck
  • Genome-wide CRISPR screenings identified SMCHD1 as a host-restricting factor for AAV transduction.
Genome-wide CRISPR screenings identified SMCHD1 as a host-restricting factor for AAV transduction.
PLoS pathogens (2024-07-08)
Chenlu Wang, Yu Liu, Jingfei Xiong, Kun Xie, Tianshu Wang, Yu Hu, Huancheng Fu, Baiquan Zhang, Xiaochao Huang, Hui Bao, Haoyang Cai, Biao Dong, Zhonghan Li
ABSTRACT
AAV-mediated gene therapy typically requires a high dose of viral transduction, risking acute immune responses and patient safety, part of which is due to limited understanding of the host-viral interactions, especially post-transduction viral genome processing. Here, through a genome-wide CRISPR screen, we identified SMCHD1 (Structural Maintenance of Chromosomes Hinge Domain 1), an epigenetic modifier, as a critical broad-spectrum restricting host factor for post-entry AAV transgene expression. SMCHD1 knock-down by RNAi and CRISPRi or knock-out by CRISPR all resulted in significantly enhanced transgene expression across multiple viral serotypes, as well as for both single-strand and self-complementary AAV genome types. Mechanistically, upon viral transduction, SMCHD1 effectively repressed AAV transcription by the formation of an LRIF1-HP1-containing protein complex and directly binding with the AAV genome to maintain a heterochromatin-like state. SMCHD1-KO or LRIF1-KD could disrupt such a complex and thus result in AAV transcriptional activation. Together, our results highlight the host factor-induced chromatin remodeling as a critical inhibitory mechanism for AAV transduction and may shed light on further improvement in AAV-based gene therapy.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Ligand-dependent nuclear receptor-interacting factor 1 Antibody, 0.5 mg/mL, from rabbit