Merck
  • Ex Vivo Functional Assay for Evaluating Treatment Response in Tumor Tissue of Head and Neck Squamous Cell Carcinoma.
Ex Vivo Functional Assay for Evaluating Treatment Response in Tumor Tissue of Head and Neck Squamous Cell Carcinoma.
Cancers (2023-01-22)
Marta E Capala, Katrin S Pachler, Iris Lauwers, Maarten A de Korte, Nicole S Verkaik, Hetty Mast, Brend P Jonker, Aniel Sewnaik, Jose A Hardillo, Stijn Keereweer, Dominiek Monserez, Senada Koljenovic, Bianca Mostert, Gerda M Verduijn, Steven Petit, Dik C van Gent
ABSTRACT
Head and neck squamous cell carcinoma (HNSCC) displays a large heterogeneity in treatment response, and consequently in patient prognosis. Despite extensive efforts, no clinically validated model is available to predict tumor response. Here we describe a functional test for predicting tumor response to radiation and chemotherapy on the level of the individual patient. Resection material of 17 primary HNSCC patients was cultured ex vivo, irradiated or cisplatin-treated, after which the effect on tumor cell vitality was analyzed several days after treatment. Ionizing radiation (IR) affected tumor cell growth and viability with a clear dose-response relationship, and marked heterogeneity between tumors was observed. After a single dose of 5Gy, proliferation in IR-sensitive tumors dropped below 30% of the untreated level, while IR-resistant tumors maintained at least 60% of proliferation. IR-sensitive tumors showed on average a twofold increase in apoptosis, as well as an increased number and size of DNA damage foci after treatment. No differences in the homologous recombination (HR) proficiency between IR-sensitive and -resistant tumors were detected. Cisplatin caused a decrease in proliferation, as well as induction of apoptosis, again with marked variation between the samples. Our functional ex vivo assay discriminated between IR-sensitive and IR-resistant HNSCC tumors, and may also be suitable for predicting response to cisplatin. Its predictive value is currently under investigation in a prospective clinical study.
Sigma-Aldrich
Fibroblast Growth Factor-Basic, FGF-Basic, from human, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture