Merck
  • Melatonin accelerates the developmental competence and telomere elongation in ovine SCNT embryos.
Melatonin accelerates the developmental competence and telomere elongation in ovine SCNT embryos.
PloS one (2022-07-22)
Parisa Nadri, Saeid Ansari-Mahyari, Farnoosh Jafarpour, Amir Hossein Mahdavi, Nima Tanhaei Vash, Liana Lachinani, Kianoush Dormiani, Mohammad Hossein Nasr-Esfahani
ABSTRACT
SCNT embryos suffer from poor developmental competence (both in vitro and in vivo) due to various defects such as oxidative stress, incomplete epigenetic reprogramming, and flaws in telomere rejuvenation. It is very promising to ameliorate all these defects in SCNT embryos by supplementing the culture medium with a single compound. It has been demonstrated that melatonin, as a multitasking molecule, can improve the development of SCNT embryos, but its function during ovine SCNT embryos is unclear. We observed that supplementation of embryonic culture medium with 10 nM melatonin for 7 days accelerated the rate of blastocyst formation in ovine SCNT embryos. In addition, the quality of blastocysts increased in the melatonin-treated group compared with the SCNT control groups in terms of ICM, TE, total cell number, and mRNA expression of NANOG. Mechanistic studies in this study revealed that the melatonin-treated group had significantly lower ROS level, apoptotic cell ratio, and mRNA expression of CASPASE-3 and BAX/BCL2 ratio. In addition, melatonin promotes mitochondrial membrane potential and autophagy status (higher number of LC3B dots). Our results indicate that melatonin decreased the global level of 5mC and increased the level of H3K9ac in the treated blastocyst group compared with the blastocysts in the control group. More importantly, we demonstrated for the first time that melatonin treatment promoted telomere elongation in ovine SCNT embryos. This result offers the possibility of better development of ovine SCNT embryos after implantation. We concluded that melatonin can accelerate the reprogramming of telomere length in sheep SCNT embryos, in addition to its various beneficial effects such as increasing antioxidant capacity, reducing DNA damage, and improving the quality of derived blastocysts, all of which led to a higher in vitro development rate.
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
Anti-acetyl-Histone H3 (Ac-Lys9) antibody, Mouse monoclonal, clone AH3-120, purified from hybridoma cell culture