Merck
  • KSK19 - Novel histamine H3 receptor ligand reduces body weight in diet induced obese mice.
KSK19 - Novel histamine H3 receptor ligand reduces body weight in diet induced obese mice.
Biochemical pharmacology (2019-07-12)
Magdalena Kotańska, Kamil Mika, Karolina Reguła, Katarzyna Szczepańska, Małgorzata Szafarz, Marek Bednarski, Agnieszka Olejarz-Maciej, Katarzyna Nowak, Gniewomir Latacz, Szczepan Mogilski, Kamil J Kuder, Katarzyna Kieć-Kononowicz, Jacek Sapa
ABSTRACT
Histamine H3 receptors ligands act anorectic by blocking the H3 autoreceptors in the CNS, that results in increased synthesis and disinhibition of histamine release. Histamine further influencing H1 receptors participates in the leptin-dependent inhibition of food intake. It also affects the peripheral metabolism by increasing white adipose tissue lipolysis. Thus, ligands such as KSK19 with significant antagonistic properties at the H3 receptor might serve as an useful treatment for obesity. To induce obesity, female CD-1 mice were fed a high-fat diet for 14 weeks. The test compound at the doses of 10 or 15 mg/kg, i.p. was administrated for 21 days. Glucose and insulin tolerance tests was performed at the beginning of week 15. At the end of study, amount of intraperitoneal fat pads, AlAT, IL-6 and TNF-α plasma levels were determined. Animals fed with high-fat diet and treated with test compound at the dose of 15 mg/kg showed significantly less weight gain, than mice from the control group. The use of KSK19 for 21 days in obese mice also significantly improved glucose tolerance and insulin resistance. In the tested doses KSK19 did not affect locomotor activity neither in lean nor in obese mice after single i.p. administration, but spontaneous activity increased during three hour after twentieth administrations. KSK19 is a strong, selective histamine H3 receptor antagonist with a favorable influence on body weight after multiple administration at the dose of 15 mg/kg. Moreover it significantly improves glucose tolerance.
Sigma-Aldrich
(R)(−)-α-Methylhistamine dihydrochloride, solid, ≥98% (HPLC)