Merck
  • Loss of PKA regulatory subunit 1α aggravates cardiomyocyte necrosis and myocardial ischemia/reperfusion injury.
Loss of PKA regulatory subunit 1α aggravates cardiomyocyte necrosis and myocardial ischemia/reperfusion injury.
The Journal of biological chemistry (2021-06-05)
Yuening Liu, Jingrui Chen, Peng Xia, Constantine A Stratakis, Zhaokang Cheng
ABSTRACT
Reperfusion therapy, the standard treatment for acute myocardial infarction, can trigger necrotic death of cardiomyocytes and provoke ischemia/reperfusion (I/R) injury. However, signaling pathways that regulate cardiomyocyte necrosis remain largely unknown. Our recent genome-wide RNAi screen has identified a potential necrosis suppressor gene PRKAR1A, which encodes PKA regulatory subunit 1α (R1α). R1α is primarily known for regulating PKA activity by sequestering PKA catalytic subunits in the absence of cAMP. Here, we showed that depletion of R1α augmented cardiomyocyte necrosis in vitro and in vivo, resulting in exaggerated myocardial I/R injury and contractile dysfunction. Mechanistically, R1α loss downregulated the Nrf2 antioxidant transcription factor and aggravated oxidative stress following I/R. Degradation of the endogenous Nrf2 inhibitor Keap1 through p62-dependent selective autophagy was blocked by R1α depletion. Phosphorylation of p62 at Ser349 by mammalian target of rapamycin complex 1 (mTORC1), a critical step in p62-Keap1 interaction, was induced by I/R, but diminished by R1α loss. Activation of PKA by forskolin or isoproterenol almost completely abolished hydrogen-peroxide-induced p62 phosphorylation. In conclusion, R1α loss induces unrestrained PKA activation and impairs the mTORC1-p62-Keap1-Nrf2 antioxidant defense system, leading to aggravated oxidative stress, necrosis, and myocardial I/R injury. Our findings uncover a novel role of PKA in oxidative stress and necrosis, which may be exploited to develop new cardioprotective therapies.
Sigma-Aldrich
ITS Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%
Roche
Cytotoxicity Detection Kit (LDH), suitable for protein quantification, suitable for cell analysis, detection, sufficient for ≤2,000 tests
Sigma-Aldrich
Anti-Nrf2 Antibody, clone 103, clone 103, from rat