Merck
  • ABHD5 inhibits YAP-induced c-Met overexpression and colon cancer cell stemness via suppressing YAP methylation.
ABHD5 inhibits YAP-induced c-Met overexpression and colon cancer cell stemness via suppressing YAP methylation.
Nature communications (2021-11-20)
Yan Gu, Yanrong Chen, Lai Wei, Shuang Wu, Kaicheng Shen, Chengxiang Liu, Yan Dong, Yang Zhao, Yue Zhang, Chi Zhang, Wenling Zheng, Jiangyi He, Yunlong Wang, Yifei Li, Xiaoxin Zhao, Hongwei Wang, Jun Tan, Liting Wang, Qi Zhou, Ganfeng Xie, Houjie Liang, Juanjuan Ou
ABSTRACT
Cancer stemness represents a major source of development and progression of colorectal cancer (CRC). c-Met critically contributes to CRC stemness, but how c-Met is activated in CRC remains elusive. We previously identified the lipolytic factor ABHD5 as an important tumour suppressor gene in CRC. Here, we show that loss of ABHD5 promotes c-Met activation to sustain CRC stemness in a non-canonical manner. Mechanistically, we demonstrate that ABHD5 interacts in the cytoplasm with the core subunit of the SET1A methyltransferase complex, DPY30, thereby inhibiting the nuclear translocation of DPY30 and activity of SET1A. In the absence of ABHD5, DPY30 translocates to the nucleus and supports SET1A-mediated methylation of YAP and histone H3, which sequesters YAP in the nucleus and increases chromatin accessibility to synergistically promote YAP-induced transcription of c-Met, thus promoting the stemness of CRC cells. This study reveals a novel role of ABHD5 in regulating histone/non-histone methylation and CRC stemness.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Insulin human, meets USP testing specifications
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder