Merck
  • Application of a catalytic asymmetric Povarov reaction using chiral ureas to the synthesis of a tetrahydroquinoline library.
Application of a catalytic asymmetric Povarov reaction using chiral ureas to the synthesis of a tetrahydroquinoline library.
ACS combinatorial science (2012-10-24)
Baudouin Gerard, Morgan Welzel O'Shea, Etienne Donckele, Sarathy Kesavan, Lakshmi B Akella, Hao Xu, Eric N Jacobsen, Lisa A Marcaurelle
ABSTRACT
A 2328-membered library of 2,3,4-trisubstituted tetrahydroquinolines was produced using a combination of solution- and solid-phase synthesis techniques. A tetrahydroquinoline (THQ) scaffold was prepared via an asymmetric Povarov reaction using cooperative catalysis to generate three contiguous stereogenic centers. A matrix of 4 stereoisomers of the THQ scaffold was prepared to enable the development of stereo/structure-activity relationships (SSAR) upon biological testing. A sparse matrix design strategy was employed to select library members to be synthesized with the goal of generating a diverse collection of tetrahydroquinolines with physicochemical properties suitable for downstream discovery.
Product Description
BRD0539, ≥98% (HPLC)
Sigma-Aldrich
(R)-N-[(1R,2R)-2-(3-(3,5-Bis(trifluoromethyl)phenyl)ureido)cyclohexyl]-tert-butyl-sulfinamide, 96%