Merck
  • Bone marrow-derived myeloid progenitors in the leptomeninges of adult mice.
Bone marrow-derived myeloid progenitors in the leptomeninges of adult mice.
Stem cells (Dayton, Ohio) (2020-12-04)
Tobias Koeniger, Luisa Bell, Anika Mifka, Michael Enders, Valentin Hautmann, Subba Rao Mekala, Philipp Kirchner, Arif B Ekici, Christian Schulz, Philipp Wörsdörfer, Stine Mencl, Christoph Kleinschnitz, Süleyman Ergün, Stefanie Kuerten
ABSTRACT
Although the bone marrow contains most hematopoietic activity during adulthood, hematopoietic stem and progenitor cells can be recovered from various extramedullary sites. Cells with hematopoietic progenitor properties have even been reported in the adult brain under steady-state conditions, but their nature and localization remain insufficiently defined. Here, we describe a heterogeneous population of myeloid progenitors in the leptomeninges of adult C57BL/6 mice. This cell pool included common myeloid, granulocyte/macrophage, and megakaryocyte/erythrocyte progenitors. Accordingly, it gave rise to all major myelo-erythroid lineages in clonogenic culture assays. Brain-associated progenitors persisted after tissue perfusion and were partially inaccessible to intravenous antibodies, suggesting their localization behind continuous blood vessel endothelium such as the blood-arachnoid barrier. Flt3Cre lineage tracing and bone marrow transplantation showed that the precursors were derived from adult hematopoietic stem cells and were most likely continuously replaced via cell trafficking. Importantly, their occurrence was tied to the immunologic state of the central nervous system (CNS) and was diminished in the context of neuroinflammation and ischemic stroke. Our findings confirm the presence of myeloid progenitors at the meningeal border of the brain and lay the foundation to unravel their possible functions in CNS surveillance and local immune cell production.
Sigma-Aldrich
Giemsa Stain, Modified Solution, (for the staining (of cellular blood components and blood parasites))
Sigma-Aldrich
May-Grünwald solution, for the staining of blood constituents