Merck
  • Kv1.3 channel blockade alleviates cerebral ischemia/reperfusion injury by reshaping M1/M2 phenotypes and compromising the activation of NLRP3 inflammasome in microglia.
Kv1.3 channel blockade alleviates cerebral ischemia/reperfusion injury by reshaping M1/M2 phenotypes and compromising the activation of NLRP3 inflammasome in microglia.
Experimental neurology (2020-07-12)
Dai-Chao Ma, Nan-Nan Zhang, Yi-Na Zhang, Hui-Sheng Chen
ABSTRACT
After cerebral ischemia/reperfusion injury, pro-inflammatory M1-like and anti-inflammatory M2-like phenotypes of microglia are involved in neuroinflammation, in which NLRP3 inflammasome plays an essential role. Kv1.3 channel has been recognized as neuro-immunomodulatory target, but it is not clear as to its role in the neuroinflammation after cerebral ischemic injury. The current study aimed to investigate the issue. Middle cerebral artery occlusion/reperfusion (MCAO/R) model in rats and oxygen-glucose deprivation/ reoxygenation (OGD/R) in primary microglia were utilized to mimic disease state of ischemic stroke. Treatment with PAP-1, a Kv1.3 channel blocker, produced a significant improvement in neurological deficit scores and a decrease in infarct volume in MCAO/R model. An increased number of M2-like phenotypic microglia and a reduced number of M1-like phenotypic microglia were observed by immunofluorescent staining in the in vivo model, which was further validated by flow cytometry in vitro. Western blot showed that PAP-1 treatment profoundly reduced cleavage of caspase-1 and IL-1β in vivo and in vitro. Furthermore, PAP-1 administration reduced the number of NLRP3+/Iba1+ cells and NLRP3 protein levels in vivo, while reduced mRNA and protein expression levels of NLRP3 in vitro. Reduced mRNA expression levels of IL-1β in vitro and protein level of IL-1β in vivo were also observed. Taken together, our findings suggested that Kv1.3 channel blockade effectively alleviated cerebral ischemic injury, possibly by reshaping microglial phenotypic response from M1 towards M2, compromising the activation of NLRP3 inflammasome in microglia, and inhibiting release of IL-1β.
Sigma-Aldrich
5-(4-Phenoxybutoxy)psoralen, ≥98% (HPLC), solid
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
EN300-1215608-50MG50 mg
Ships in 14 days - from Aldrich Partner.
DKK 3,252.75
EN300-1215608-100MG100 mg
Ships in 14 days - from Aldrich Partner.
DKK 4,666.50
EN300-1215608-250MG250 mg
Ships in 14 days - from Aldrich Partner.
DKK 6,519.00
EN300-1215608-500MG500 mg
Ships in 14 days - from Aldrich Partner.
DKK 10,058.25
EN300-1215608-1G1 g
Ships in 14 days - from Aldrich Partner.
DKK 12,807.75
EN300-1215608-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
DKK 24,771.00
EN300-1215608-5G5 g
Ships in 14 days - from Aldrich Partner.
DKK 36,490.50
EN300-1215608-10G10 g
Ships in 14 days - from Aldrich Partner.
DKK 53,952.75
To order products, please contact your local dealer.