Merck
  • Predominant cleavage of proteins N-terminal to serines and threonines using scandium(III) triflate.
Predominant cleavage of proteins N-terminal to serines and threonines using scandium(III) triflate.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2019-11-02)
Christian J Koehler, Bernd Thiede
ABSTRACT
Proteolytic digestion prior to LC-MS analysis is a key step for the identification of proteins. Digestion of proteins is typically performed with trypsin, but certain proteins or important protein sequence regions might be missed using this endoproteinase. Only few alternative endoproteinases are available and chemical cleavage of proteins is rarely used. Recently, it has been reported that some metal complexes can act as artificial proteases. In particular, the Lewis acid scandium(III) triflate has been shown to catalyze the cleavage of peptide bonds to serine and threonine residues. Therefore, we investigated if this compound can also be used for the cleavage of proteins. For this purpose, several single proteins, the 20S immune-proteasome (17 proteins), and the Universal Proteomics Standard UPS1 (48 proteins) were analyzed by MALDI-MS and/or LC-MS. A high cleavage specificity N-terminal to serine and threonine residues was observed, but also additional peptides with deviating cleavage specificity were found. Scandium(III) triflate can be a useful tool in protein analysis as no other reagent has been reported yet which showed cleavage specificity within proteins to serines and threonines.
Sigma-Aldrich
Universal Proteomics Standard Set, Protein Mass Spectrometry Calibration Standard