Merck
  • Kinetic characterization of VIM-7, a divergent member of the VIM metallo-beta-lactamase family.
Kinetic characterization of VIM-7, a divergent member of the VIM metallo-beta-lactamase family.
Antimicrobial agents and chemotherapy (2008-06-19)
Ørjan Samuelsen, Mariana Castanheira, Timothy R Walsh, James Spencer
ABSTRACT
Purified recombinant VIM-7 possesses efficient penicillinase and carbapenemase activities comparable to those of VIM-2. Cephalosporinase activity was variable and generally lower than those of VIM-1 and VIM-2. A homology model suggests that the VIM-7 Tyr-218 Phe substitution may be responsible for the reduced catalytic efficiency against certain cephalosporins, including ceftazidime and cefepime.
Sigma-Aldrich
Terrific Broth, Liquid microbial growth medium
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered