Merck
  • MiT/TFE factors control ER-phagy via transcriptional regulation of FAM134B.
MiT/TFE factors control ER-phagy via transcriptional regulation of FAM134B.
The EMBO journal (2020-07-28)
Laura Cinque, Chiara De Leonibus, Maria Iavazzo, Natalie Krahmer, Daniela Intartaglia, Francesco Giuseppe Salierno, Rossella De Cegli, Chiara Di Malta, Maria Svelto, Carmela Lanzara, Marianna Maddaluno, Luca Giorgio Wanderlingh, Antje K Huebner, Marcella Cesana, Florian Bonn, Elena Polishchuk, Christian A Hübner, Ivan Conte, Ivan Dikic, Matthias Mann, Andrea Ballabio, Francesca Sacco, Paolo Grumati, Carmine Settembre
ABSTRACT
Lysosomal degradation of the endoplasmic reticulum (ER) via autophagy (ER-phagy) is emerging as a critical regulator of cell homeostasis and function. The recent identification of ER-phagy receptors has shed light on the molecular mechanisms underlining this process. However, the signaling pathways regulating ER-phagy in response to cellular needs are still largely unknown. We found that the nutrient responsive transcription factors TFEB and TFE3-master regulators of lysosomal biogenesis and autophagy-control ER-phagy by inducing the expression of the ER-phagy receptor FAM134B. The TFEB/TFE3-FAM134B axis promotes ER-phagy activation upon prolonged starvation. In addition, this pathway is activated in chondrocytes by FGF signaling, a critical regulator of skeletal growth. FGF signaling induces JNK-dependent proteasomal degradation of the insulin receptor substrate 1 (IRS1), which in turn inhibits the PI3K-PKB/Akt-mTORC1 pathway and promotes TFEB/TFE3 nuclear translocation and enhances FAM134B transcription. Notably, FAM134B is required for protein secretion in chondrocytes, and cartilage growth and bone mineralization in medaka fish. This study identifies a new signaling pathway that allows ER-phagy to respond to both metabolic and developmental cues.
Sigma-Aldrich
Anti-phospho-IRS1 (Ser307 mouse/ Ser312 human) Antibody, clone 24.6.2, clone 24.6.2, from mouse
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Anti-FAM134B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody