Merck
  • Wall teichoic acid protects Staphylococcus aureus against antimicrobial fatty acids from human skin.
Wall teichoic acid protects Staphylococcus aureus against antimicrobial fatty acids from human skin.
Journal of bacteriology (2009-05-12)
Thomas Kohler, Christopher Weidenmaier, Andreas Peschel
ABSTRACT
Skin-colonizing gram-positive bacteria produce wall teichoic acids (WTAs) or related glycopolymers for unclear reasons. Using a WTA-deficient Staphylococcus aureus mutant, we demonstrated that WTA confers resistance to antimicrobial fatty acids from human sebaceous glands by preventing fatty acid binding. Thus, WTA is probably important for bacterial skin colonization.
Product Description
Millipore
Mueller Hinton Broth 2, NutriSelect® Plus, Cation-adjusted, suitable for microbiology
Millipore
Mueller Hinton Broth, suitable for microbiology, NutriSelect® Plus