Merck
  • Clinical implications of insulin-like growth factor 1 system in early-stage cervical cancer.
Clinical implications of insulin-like growth factor 1 system in early-stage cervical cancer.
British journal of cancer (2008-09-11)
Y-F Huang, M-R Shen, K-F Hsu, Y-M Cheng, C-Y Chou
ABSTRACT
This study was aimed to identify the expression and the correlation of insulin-like growth factor-1 (IGF-1) system and their prognostic impacts in cervical cancer. Seventy-two patients with early-stage cervical cancer were eligible. We obtained the serum levels of total IGF-1 and IGF binding protein-3 (IGFBP-3) by enzyme-linked immunosorbent assay and the expression of IGF-1 receptor (IGF-1R) in cancerous tissue by immuno-fluorescent (IF) stains. The 5-year recurrence-free and overall survival rates were significantly lower (P=0.003 and P=0.01, respectively) among patients with high-grade expression of tissue IGF-1R, compared with those with low-grade expression. After adjustment for other factors, preoperative serum total IGF-1 or IGFBP-3 levels failed to predict cancer death and recurrence. High-grade expression of IGF-1R and elevated preoperative squamous cell carcinoma antigen level were independent predictors of both death and recurrence, and combination of both factors could further help identify the subgroup of patients at higher death risk. The IF staining indicates the colocalisation of IGF-1 and IGF-1R in the cancerous tissues, whereas the IGF-1R expression is not correlated with circulating levels of IGF-1 or IGFBP-3. In early-stage cervical cancer, IGF-1 system may have a paracrine or autocrine function and the adverse impacts on prognosis by IGF-1R overexpression are implicated.
Sigma-Aldrich
Anti-Insulin-like Growth Factor-1 Receptor Antibody, α-subunit, clone 24-31, clone 24-31, Chemicon®, from mouse
Sigma-Aldrich
Anti-Insulin-like Growth Factor-I Antibody, clone M23, clone M23, Chemicon®, from mouse