Merck
  • Sonochemical synthesis of novel C60 fullerene 1,4-oxathiane derivative through the intermediate fullerene radical anion.
Sonochemical synthesis of novel C60 fullerene 1,4-oxathiane derivative through the intermediate fullerene radical anion.
Ultrasonics sonochemistry (2020-05-18)
Zemfira S Kinzyabaeva, Denis Sh Sabirov
ABSTRACT
We have discovered a selective and efficient method for the synthesis of previously unknown 1,9-(1',4'-oxathiano)-1,9-dihydro-(C60-Ih)[5,6]fullerene, a compound with the direct attachment of the sulfur atom to the fullerene core. The method is based on the reaction of C60 with 1,2-hydroxythiols in the presence of inorganic bases in air under ultrasonication. The significance of ultrasound has been exemplified with the comparative conventional methods. The title compound has been identified with mass-(MALDI TOF/TOF), one- and two-dimensional NMR 1H and 13C (COSY, HSQC, HMBS), IR, UV-Vis spectroscopic techniques. Using the direct EPR method, we have detected radical anion C60- (g = 2.0046 and ΔH = 2 G) as a key reaction intermediate of the sonochemical reaction. Based on the experimental results and quantum chemical calculations, we have proposed a mechanism for the conversion of C60 and 2-mercaptoethanol to the C60-1,4-oxathiane adduct.
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Pricing and availability is not currently available.
Sigma-Aldrich
1-Mercapto-2-propanol, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Mercapto-3-butanol, mixture of isomers, ≥97%, FG
Pricing and availability is not currently available.