Merck
  • Blockade of β-Catenin-Induced CCL28 Suppresses Gastric Cancer Progression via Inhibition of Treg Cell Infiltration.
Blockade of β-Catenin-Induced CCL28 Suppresses Gastric Cancer Progression via Inhibition of Treg Cell Infiltration.
Cancer research (2020-03-12)
Lu Ji, Wei Qian, Liming Gui, Zhongzhong Ji, Pan Yin, Guan Ning Lin, You Wang, Bin Ma, Wei-Qiang Gao
ABSTRACT
Dysregulation of Wnt/β-catenin signaling is frequently observed in human gastric cancer. Elucidation of the tumor immune microenvironment is essential for understanding tumorigenesis and for the development of immunotherapeutic strategies. However, it remains unclear how β-catenin signaling regulates the tumor immune microenvironment in the stomach. Here, we identify CCL28 as a direct transcriptional target gene of β-catenin/T-cell factor (TCF). Protein levels of β-catenin and CCL28 positively correlated in human gastric adenocarcinoma. β-Catenin-activated CCL28 recruited regulatory T (Treg) cells in a transwell migration assay. In a clinically relevant mouse gastric cancer model established by Helicobacter (H.) felis infection and N-methyl-N-nitrosourea (MNU) treatment, inhibition of β-catenin/TCF activity by a pharmacologic inhibitor iCRT14 suppressed CCL28 expression and Treg cell infiltration in the stomach. Moreover, an anti-CCL28 antibody attenuated Treg cell infiltration and tumor progression in H. felis/MNU mouse models. Diphtheria toxin-induced Treg cell ablation restrained gastric cancer progression in H. felis/MNU-treated DEREG (Foxp3-DTR) mice, clarifying the tumor-promoting role of Treg cells. Thus, the β-catenin-CCL28-Treg cell axis may serve as an important mechanism for immunosuppression of the stomach tumor microenvironment. Our findings reveal an immunoregulatory role of β-catenin signaling in stomach tumors and highlight the therapeutic potential of CCL28 blockade for the treatment of gastric cancer. SIGNIFICANCE: These findings demonstrate an immunosuppressive role of tumor-intrinsic β-catenin signaling and the therapeutic potential of CCL28 blockade in gastric cancer.
Sigma-Aldrich
Human CCL28 / VIC ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
iCRT14, ≥98% (HPLC)