Merck
  • XIST lost induces ovarian cancer stem cells to acquire taxol resistance via a KMT2C-dependent way.
XIST lost induces ovarian cancer stem cells to acquire taxol resistance via a KMT2C-dependent way.
Cancer cell international (2020-09-19)
Ruili Huang, Lijuan Zhu, Yali Zhang
ABSTRACT
The expression levels of long non-coding RNA XIST are significantly associated with paclitaxel (Pac) sensitivity in ovarian cancer, but the mechanism of action remains unclear. Therefore, this experimental design was based on lncRNA XIST analysis to regulate the effect of XIST on the tumor stem cell and paclitaxel sensitivity in ovarian cancer. Sphere assay and fluorescence activated cell sorting (FACS) were used to determine the expression levels of XIST and sensitivity to paclitaxel treatment. The effect of the proliferation was detected by MTT assay. Target gene prediction and screening, luciferase reporter assays were used to validate downstream target genes for lncRNA XIS and KMT2C. The expression of KMT2C was detected by RT-qPCR and Western blotting. RT-qPCR was used to detect the expression of cancer stem cell-associated genes SOX2, OCT4 and Nanog. The tumor changes in mice were detected by in vivo experiments in nude mice. There was an inverse correlation between the expression of XIST and cancer stem cell (CD44 + /CD24-) population. XIST promoted methylation of histone H3 methylation at lysine 4 by enhancing the stability of lysine (K)-specific methyltransferase 2C (KMT2C) mRNA. XIST acted on the stability of KMT2C mRNA by directly targeting miR-93-5p. Overexpression of miR-93-5p can reverse the XIST overexpression-induced KMT2C decrease and sphere number increase. Overexpression of KMT2C inhibited XIST silencing-induced proliferation of cancer stem cells, and KMT2C was able to mediate paclitaxel resistance induced by XIST in ovarian cancer. The study found that XIST can affect the expression of KMT2C in the ovarian cancer via targeting miR-93-5p. XIST promoted the sensitivity of ovarian cancer stem cells to paclitaxel in a KMT2C-dependent manner.
Sigma-Aldrich
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Monoclonal Anti-AGO2 antibody produced in rat, ~1.5 mg/mL, clone 11A9, purified immunoglobulin