Merck
  • Nuclear inclusions of pathogenic ataxin-1 induce oxidative stress and perturb the protein synthesis machinery.
Nuclear inclusions of pathogenic ataxin-1 induce oxidative stress and perturb the protein synthesis machinery.
Redox biology (2020-03-08)
Stamatia Laidou, Gregorio Alanis-Lobato, Jan Pribyl, Tamás Raskó, Boris Tichy, Kamil Mikulasek, Maria Tsagiopoulou, Jan Oppelt, Georgia Kastrinaki, Maria Lefaki, Manvendra Singh, Annika Zink, Niki Chondrogianni, Fotis Psomopoulos, Alessandro Prigione, Zoltán Ivics, Sarka Pospisilova, Petr Skladal, Zsuzsanna Izsvák, Miguel A Andrade-Navarro, Spyros Petrakis
ABSTRACT
Spinocerebellar ataxia type-1 (SCA1) is caused by an abnormally expanded polyglutamine (polyQ) tract in ataxin-1. These expansions are responsible for protein misfolding and self-assembly into intranuclear inclusion bodies (IIBs) that are somehow linked to neuronal death. However, owing to lack of a suitable cellular model, the downstream consequences of IIB formation are yet to be resolved. Here, we describe a nuclear protein aggregation model of pathogenic human ataxin-1 and characterize IIB effects. Using an inducible Sleeping Beauty transposon system, we overexpressed the ATXN1(Q82) gene in human mesenchymal stem cells that are resistant to the early cytotoxic effects caused by the expression of the mutant protein. We characterized the structure and the protein composition of insoluble polyQ IIBs which gradually occupy the nuclei and are responsible for the generation of reactive oxygen species. In response to their formation, our transcriptome analysis reveals a cerebellum-specific perturbed protein interaction network, primarily affecting protein synthesis. We propose that insoluble polyQ IIBs cause oxidative and nucleolar stress and affect the assembly of the ribosome by capturing or down-regulating essential components. The inducible cell system can be utilized to decipher the cellular consequences of polyQ protein aggregation. Our strategy provides a broadly applicable methodology for studying polyQ diseases.
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, original GFP only, clone 264-449-2, clone 264-449-2, Chemicon®, from mouse