Merck
Age-dependent increase in Thy-1 protein in the rat supraoptic nucleus.
Heliyon (2020-03-18)
Jason M Askvig, Talia S Dalzell, Nadia Toumeh, Phillip T Kuball, Sara T Whiteman, Erik W Bye, Marissa J Andersen, Michael G McCarthy, Riley E Irmen, Sydney H Bexell, Molly M Benolken, Brooke L Maruska, Shelby E Nordmann
ABSTRACT
Mature mammalian CNS neurons often do not recover successfully following injury. To this point, unilateral lesion of the hypothalamo-neurohypophysial tract results in collateral sprouting from uninjured axons of the supraoptic nucleus (SON) in 35-day-old but not in 125-day-old rats. Thus, it appears that there are age-related changes within the SON that preclude the older rat from recovering following axotomy. We hypothesize that the intrinsic capacity for axon reorganization may depend, in part, on age-related alterations in cell adhesion molecules that allow normal astrocyte-neuron interactions in the SON. In support of our hypothesis, numerous reports have shown that Thy-1 is increased in neurons at the cessation of axon outgrowth. Therefore, we compared protein levels of Thy-1 and the Thy-1 interacting integrin subunits, alpha-v (αv), beta-3 (ß3), and beta-5 (ß5), in 35- and 125-day-old SON using western blot analysis. Our results demonstrated that there was significantly more Thy-1 protein in the 125-day-old SON compared to 35-day-old SON, but no change in the protein levels of the integrin subunits. Furthermore, we localized Thy-1-, αv integrin-, ß3 integrin-, and ß5 integrin-immunoreactivity to both neurons and astrocytes in the SON. Altogether, our results suggest that the observed increase in Thy-1 protein levels in the SON with age may contribute to an environment that prevents collateral axonal sprouting in the SON of the 125-day-old rat.
Sigma-Aldrich
Anti-Thy-1.1 Antibody, clone OX-7, clone OX-7, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution