Merck
  • Profiling and structural analysis of cardenolides in two species of Digitalis using liquid chromatography coupled with high-resolution mass spectrometry.
Profiling and structural analysis of cardenolides in two species of Digitalis using liquid chromatography coupled with high-resolution mass spectrometry.
Journal of chromatography. A (2020-02-10)
Baradwaj Gopal Ravi, Mary Grace E Guardian, Rebecca Dickman, Zhen Q Wang
ABSTRACT
Plants of the Digitalis genus contain a cocktail of cardenolides commonly prescribed to treat heart failure. Cardenolides in Digitalis extracts have been conventionally quantified by high-performance liquid chromatography yet the lack of structural information compounded with possible co-eluents renders this method insufficient for analyzing cardenolides in plants. The goal of this work is to structurally characterize cardiac glycosides in fresh-leaf extracts using liquid chromatography coupled with tandem mass spectrometry (LC/MS/MS) that provides measured accurate mass. Fragmentation of cardenolides is featured by sequential loss of sugar units while the steroid aglycone moieties undergo stepwise elimination of hydroxyl groups, which distinguishes different aglycones. Using a reverse-phase LC column, the sequence of elution follows: diginatigenin→digoxigenin→gitoxigenin→gitaloxigenin→digitoxigenin for cardenolides with the same sugar units but different aglycones. A linear range of 0.8-500 ng ml-1 has been achieved for digoxigenin, β-acetyldigoxin, and digitoxigenin with limits of detection ranging from 0.09 to 0.45 ngml-1. A total of seventeen cardenolides have been detected with lanatoside A, C, and E as major cardenolides in Digitalis lanata while seven have been found in Digitalis purpurea including purpurea glycoside A, B, and E. Surprisingly, glucodigifucoside in D. lanata and verodoxin and digitoxigenin fucoside in D. purpurea have also been found as major cardenolides. As the first MS/MS-based method developed for analyzing cardenolides in plant extracts, this method serves as a foundation for complete identification and accurate quantification of cardiac glycosides, a necessary step towards understanding the biosynthesis of cardenolide in plants.
Sigma-Aldrich
Digitoxigenin
Sigma-Aldrich
Lanatoside C, ≥97% (TLC)