Merck
Fear renewal requires nitric oxide signaling in the lateral amygdala.
Biochemical and biophysical research communications (2019-12-14)
Sukwoon Song, Junghwa Lee, Sewon Park, Sukwoo Choi
ABSTRACT
Fear renewal is defined as return of the conditioned fear responses after extinction when a conditioned stimulus (CS) is given outside of the extinction context. Previously, we have suggested that extinction induces S-nitrosylation of GluA1 in the lateral amygdala (LA), and that the extinction-induced S-nitrosylation of GluA1 lowers the threshold of GluA1 phosphorylation (at Ser 831) which is required for fear renewal. This fits nicely with the fact that fear renewal is induced by weak stimuli. However, it has not been tested whether S-nitrosylation of GluA1 in the LA is indeed required for fear renewal. In the present study, we used three different chemicals to impede protein S-nitrosylation via distinct mechanisms. Fear renewal was inhibited by microinjection of 7-Nitroindazole (nNOS inhibitor), and ZL006 (a blocker of PSD-95-nNOS interaction) before fear renewal. Furthermore, fear renewal was also attenuated by microinjection of a strong antioxidant (N-acetyl cysteine), which scavenges reactive oxygen including nitric oxide, into the LA before each extinction training. These findings suggest that protein S-nitrosylation is required for fear renewal.
Sigma-Aldrich
ZL006, ≥98% (HPLC)
Sigma-Aldrich
7-Nitroindazole, ≥98%
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)