Merck
  • Transferrin plays a central role in coagulation balance by interacting with clotting factors.
Transferrin plays a central role in coagulation balance by interacting with clotting factors.
Cell research (2019-12-08)
Xiaopeng Tang, Zhiye Zhang, Mingqian Fang, Yajun Han, Gan Wang, Sheng Wang, Min Xue, Yaxiong Li, Li Zhang, Jian Wu, Biqing Yang, James Mwangi, Qiumin Lu, Xiaoping Du, Ren Lai
ABSTRACT
Coagulation balance is maintained through fine-tuned interactions among clotting factors, whose physiological concentrations vary substantially. In particular, the concentrations of coagulation proteases (pM to nM) are much lower than their natural inactivator antithrombin (AT, ~ 3 μM), suggesting the existence of other coordinators. In the current study, we found that transferrin (normal plasma concentration ~40 μM) interacts with fibrinogen, thrombin, factor XIIa (FXIIa), and AT with different affinity to maintain coagulation balance. Normally, transferrin is sequestered by binding with fibrinogen (normal plasma concentration ~10 μM) at a molar ratio of 4:1. In atherosclerosis, abnormally up-regulated transferrin interacts with and potentiates thrombin/FXIIa and blocks AT's inactivation effect on coagulation proteases by binding to AT, thus inducing hypercoagulability. In the mouse model, transferrin overexpression aggravated atherosclerosis, whereas transferrin inhibition via shRNA knockdown or treatment with anti-transferrin antibody or designed peptides interfering with transferrin-thrombin/FXIIa interactions alleviated atherosclerosis. Collectively, these findings identify that transferrin is an important clotting regulator and an adjuster in the maintenance of coagulation balance and modifies the coagulation cascade.
Sigma-Aldrich
Human TF / Serotransferrin ELISA Kit
Sigma-Aldrich
Antithrombin III from human plasma, lyophilized powder, ≥95% (SDS-PAGE)
Sigma-Aldrich
RNAlater®, Stabilize and protect RNA with immediate RNase inactivation
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)