Merck
  • The superiority of conditioned medium derived from rapidly expanded mesenchymal stem cells for neural repair.
The superiority of conditioned medium derived from rapidly expanded mesenchymal stem cells for neural repair.
Stem cell research & therapy (2019-12-18)
Ya-Tzu Chen, May-Jywan Tsai, Nini Hsieh, Ming-Jei Lo, Meng-Jen Lee, Henrich Cheng, Wen-Cheng Huang
ABSTRACT
Spinal cord injury (SCI) is a complex and severe neurological condition. Mesenchymal stem cells (MSCs) and their secreted factors show promising potential for regenerative medicine. Many studies have investigated MSC expansion efficacy of all kinds of culture medium formulations, such as growth factor-supplemented or xeno-free medium. However, very few studies have focused on the potential of human MSC (hMSC) culture medium formulations for injured spinal cord repair. In this study, we investigated the effect of hMSC-conditioned medium supplemented with bFGF, EGF, and patient plasma, namely, neural regeneration laboratory medium (NRLM), on SCI in vitro and in vivo. Commercial and patient bone marrow hMSCs were obtained for cultivation in standard medium and NRLM separately. Several characteristics, including CD marker expression, differentiation, and growth curves, were compared between MSCs cultured in standard medium and NRLM. Additionally, we investigated the effect of the conditioned medium (referred to as NRLM-CM) on neural repair, including inflammation inhibition, neurite regeneration, and spinal cord injury (SCI), and used a coculture system to detect the neural repair function of NRLM-MSCs. Compared to standard culture medium, NRLM-CM had superior in inflammation reduction and neurite regeneration effects in vitro and improved functional restoration in SCI rats in vivo. In comparison with standard culture medium MSCs, NRLM-MSCs proliferated faster regardless of the age of the donor. NRLM-MSCs also showed increased adipose differentiative potential and reduced CD90 expression. Both types of hMSC CM effectively enhanced injured neurite outgrowth and protected against H2O2 toxicity in spinal cord neuron cultures. Cytokine arrays performed in hMSC-CM further revealed the presence of at least 120 proteins. Among these proteins, 6 demonstrated significantly increased expression in NRLM-CM: adiponectin (Acrp30), angiogenin (ANG), HGF, NAP-2, uPAR, and IGFBP2. The NRLM culture system provides rapid expansion effects and functional hMSCs. The superiority of the derived conditioned medium on neural repair shows potential for future clinical applications.
Sigma-Aldrich
Anti-BrdU Antibody, clone IIB5, clone IIB5, Chemicon®, from mouse
Sigma-Aldrich
Mesenchymal Adipogenesis Kit, Mesenchymal Stem Cell Adipogenesis Kit typically obtains 30% mature adipocytes from the rat bone marrow derived mesenchymal stem cells.