Merck
  • Laminin-binding integrins induce Dll4 expression and Notch signaling in endothelial cells.
Laminin-binding integrins induce Dll4 expression and Notch signaling in endothelial cells.
Circulation research (2011-04-09)
Soline Estrach, Laurence Cailleteau, Claudio Areias Franco, Holger Gerhardt, Caroline Stefani, Emmanuel Lemichez, Laurent Gagnoux-Palacios, Guerrino Meneguzzi, Amel Mettouchi
ABSTRACT
Integrins play a crucial role in controlling endothelial cell proliferation and migration during angiogenesis. The Delta-like 4 (Dll4)/Notch pathway establishes an adequate ratio between stalk and tip cell populations by restricting tip cell formation through "lateral inhibition" in response to a vascular endothelial growth factor gradient. Because angiogenesis requires a tight coordination of these cellular processes, we hypothesized that adhesion, vascular endothelial growth factor, and Notch signaling pathways are interconnected. This study was aimed at characterizing the cross-talk between integrin and Notch signaling in endothelial cells. Adhesion of primary human endothelial cells to laminin-111 triggers Dll4 expression, leading to subsequent Notch pathway activation. SiRNA-mediated knockdown of α2β1 and α6β1 integrins abolishes Dll4 induction, which discloses a selective integrin signaling acting upstream of Notch pathway. The increase in Foxc2 transcription, triggered by α2β1 binding to laminin, is required but not sufficient per se for Dll4 expression. Furthermore, vascular endothelial growth factor stimulates laminin γ1 deposition, which leads to integrin signaling and Dll4 induction. Interestingly, loss of integrins α2 or α6 mimics the effects of Dll4 silencing and induces excessive network branching in an in vitro sprouting angiogenesis assay on three-dimensional matrigel. We show that, in endothelial cells, ligation of α2β1 and α6β1 integrins induces the Notch pathway, and we disclose a novel role of basement membrane proteins in the processes controlling tip vs stalk cell selection.
Sigma-Aldrich
Anti-Integrin α2 Antibody, serum, Chemicon®
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid