Merck
Preparation of amino acid mixtures for cell-free expression systems.
BioTechniques (2015-01-22)
Filippo Caschera, Vincent Noireaux
ABSTRACT
Here we present a procedure for preparing amino acid mixtures--having both the desired composition and a physiological pH--at high concentrations for cell-free expression systems. Up to 2.1 mg/mL of active protein was synthesized in batch mode reactions with an all Escherichia coli cell-free expression system. Our method is fast, easy to execute, and economically advantageous compared to expensive commercial kits, making it useful for high-throughput experiments, incorporation of nonstandard amino acids, and cell-free metabolic engineering.
Sigma-Aldrich
Cell Free Amino Acid Mixture-15N, 5-100 mM in water, 98 atom % 15N
Sigma-Aldrich
Cell Free Amino Acid Mixture-13C, 15N, D, 2-100 mM in water, 98 atom % 13C, 97 atom % D, 98 atom % 15N
Sigma-Aldrich
Cell Free Amino Acid Mixture - 13C,15N, 5-100 mM in water, 98 atom % 15N, 98 atom % 13C