Merck
  • Anti-tumor activity of N-hydroxy-7-(2-naphthylthio) heptanomide, a novel histone deacetylase inhibitor.
Anti-tumor activity of N-hydroxy-7-(2-naphthylthio) heptanomide, a novel histone deacetylase inhibitor.
Biochemical and biophysical research communications (2007-03-14)
Dong Hoon Kim, Jiyong Lee, Kyung Noo Kim, Hye Jin Kim, Hei Cheul Jeung, Hyun Cheol Chung, Ho Jeong Kwon
ABSTRACT
Histone deacetylase (HDAC), a key enzyme in gene expression and carcinogenesis, is considered an attractive target molecule for cancer therapy. Here, we report a new synthetic small molecule, N-hydroxy-7-(2-naphthylthio) heptanomide (HNHA), as a HDAC inhibitor with anti-tumor activity both in vitro and in vivo. The compound inhibited HDAC enzyme activity as well as proliferation of human fibrosarcoma cells (HT1080) in vitro. Treatment of cells with HNHA elicited histone hyperacetylation leading to an up-regulation of p21 transcription, cell cycle arrest, and an inhibition of HT1080 cell invasion. Moreover, HNHA effectively inhibited the growth of tumor tissue in a mouse xenograph assay in vivo. Together, these data demonstrate that this novel HDAC inhibitor could be developed as a potential anti-tumor agent targeting HDAC.
Sigma-Aldrich
Histone Deacetylase Inhibitor VI, HNHA, The Histone Deacetylase Inhibitor VI, HNHA, also referenced under CAS 926908-04-5, controls the biological activity of Histone Deacetylase.