Merck
  • Honey-Mediated Wound Healing: H₂O₂ Entry through AQP3 Determines Extracellular Ca2+ Influx.
Honey-Mediated Wound Healing: H₂O₂ Entry through AQP3 Determines Extracellular Ca2+ Influx.
International journal of molecular sciences (2019-02-14)
Simona Martinotti, Umberto Laforenza, Mauro Patrone, Francesco Moccia, Elia Ranzato
ABSTRACT
Since Biblical times, honey has been utilized in "folk medicine", and in recent decades the positive qualities of honey have been re-discovered and are gaining acceptance. Scientific literature states that honey has been successfully utilized on infections not responding to classic antiseptic and antibiotic therapy, because of its intrinsic H₂O₂ production. In our study, we demonstrated the involvement of H₂O₂ as a main mediator of honey regenerative effects on an immortalized human keratinocyte cell line. We observed that this extracellularly released H₂O₂ could pass across the plasma membrane through a specific aquaporin (i.e., AQP3). Once in the cytoplasm H₂O₂, in turn, induces the entry of extracellular Ca2+ through Melastatin Transient Receptor Potential 2 (TRPM2) and Orai1 channels. Honey-induced extracellular Ca2+ entry results in wound healing, which is consistent with the role played by Ca2+ signaling in tissue regeneration. This is the first report showing that honey exposure increases intracellular Ca2+ concentration ([Ca2+]i), due to H₂O₂ production and redox regulation of Ca2+-permeable ion channels, opening up a new horizon for the utilization of the honey as a beneficial tool.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-AQP3 antibody produced in rabbit, 1 mg/mL, affinity isolated antibody