Merck
  • Chelation-mediated palladium(II)-catalyzed domino Heck-Mizoroki/Suzuki-Miyaura reactions using arylboronic acids: increasing scope and mechanistic understanding.
Chelation-mediated palladium(II)-catalyzed domino Heck-Mizoroki/Suzuki-Miyaura reactions using arylboronic acids: increasing scope and mechanistic understanding.
The Journal of organic chemistry (2011-03-23)
Samir Yahiaoui, Ashkan Fardost, Alejandro Trejos, Mats Larhed
ABSTRACT
A palladium(II)-catalyzed Heck-Mizoroki/Suzuki-Miyaura domino reaction involving metal coordinating dimethylaminoethyl vinyl ethers and a number of electron-rich and electron-deficient arylboronic acids has been developed. Through variation of the temperature and the concentration of the p-benzoquinone (p-Bq) ligand/reoxidant, conditions for the robust and convenient one-pot generation of diarylated-saturated ethers were identified. With the aid of coordination of the dimethylamino group to the arylpalladium intermediate, the otherwise predominant formation of the β-arylated olefin could be reversed. A reaction route involving a chelation-controlled carbopalladation, providing a p-Bq stabilized six-membered palladacycle, followed by transmetalation and reductive elimination is suggested to explain the selective formation of saturated diarylated ether products.
Sigma-Aldrich
4-Iodophenylboronic acid, ≥95.0%
Pricing and availability is not currently available.