Merck
G0S2 is an all-trans-retinoic acid target gene.
International journal of oncology (2008-07-19)
Sutisak Kitareewan, Steven Blumen, David Sekula, Reid P Bissonnette, William W Lamph, Qingping Cui, Robert Gallagher, Ethan Dmitrovsky
ABSTRACT
All-trans-retinoic acid (RA) treatment of acute promyelocytic leukemia (APL) cases expressing the t(15;17) product, PML/RARalpha, is a successful example of differentiation therapy. Uncovering RA target genes is of considerable interest in APL. This study comprehensively examines in APL cells transcriptional and post-transcriptional regulation of the novel candidate RA target gene, G0S2, the G0/G1 switch gene. Reverse transcription (RT)-polymerase chain reaction (PCR) and heteronuclear PCR assays performed +/- treatment with the protein synthesis inhibitor cycloheximide (CHX) revealed G0S2 induction within 3 h of RA-treatment. Treatment with the RNA synthesis inhibitor actinomycin D did not implicate G0S2 transcript stabilization in the RA-mediated increase of G0S2 mRNA expression. Promoter elements of G0S2 were cloned into a reporter plasmid and retinoic acid receptor (RAR) co-transfection assays confirmed transcriptional activation after RA-treatment. Consistent with G0S2 being a direct RA target gene, retinoic acid response element (RARE) half-sites were found in this promoter. Mutation of these sites blocked RA-transcriptional activation of G0S2. To extend analyses to the protein expression level, a polyclonal anti-G0S2 antibody was derived and detected murine and human G0S2 species. G0S2 protein was rapidly induced in cultured NB4-S1 human APL cells and in APL transgenic mice treated with RA. An RAR pan-antagonist confirmed dependence on RARs for this induction. That these findings are clinically relevant was shown by analyses of APL cells derived directly from patients. These leukemic cells induced both a prominent increase in the cellular differentiation marker nitrotetrazolium blue (NBT) staining and marked increase in G0S2 expression. Taken together, these findings indicate G0S2 is an RA target gene. The functional role of G0S2 in retinoid response of APL warrants further study.
Sigma-Aldrich
Monoclonal Anti-Opsin antibody produced in mouse, clone RET-P1, ascites fluid
USP
Dactinomycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture