Merck
  • Taxus globosa S. cell lines: initiation, selection and characterization in terms of growth, and of baccatin III and paclitaxel production.
Taxus globosa S. cell lines: initiation, selection and characterization in terms of growth, and of baccatin III and paclitaxel production.
Biocell : official journal of the Sociedades Latinoamericanas de Microscopia Electronica ... et. al (2010-05-29)
Dulce M Barradas-Dermitz, Patricia M Hayward-Jones, Martín Mata-Rosas, Beatriz Palmeros-Sánchez, Oscar B J Platas-Barradas, Rodolfo F Velásquez-Toledo
ABSTRACT
Of the initial six cell lines originating from explants of Taxus globosa, or Mexican yew (stem internode, leaves and meristematic tissue), three were selected for their microbial and oxidation resistance, two from leaves and the other from stem internode. A study of their behavior, both in terms of cell growth, and of baccatin III and paclitaxel production, was developed in suspension cultures with an initially standardized biomass (fresh weight 0.23 g/L) using modified Gamborg's B5 medium, and an elicitor (methyl jasmonate), on either the first or seventh day of culture, at several levels (0, 0.1, 1, 10, 100 microM). In most of the conditions used, the three cell lines showed growth associated baccatin III production. The cell line from stem internode was the highest producer of baccatin III using 1 microM elicitor, sampling at 10 days (p < or = 0.01, 6.45 mg/L). This same line also had the highest biomass production (6.85 g/L, p < or = 0.01) at 10 days of culture but at the higher elicitor concentration of 10 microM. All three cell lines did not produce paclitaxel under experimental conditions used.
Sigma-Aldrich
Gamborg′s B-5 Basal Salt Mixture, fine powder, suitable for plant cell culture
Sigma-Aldrich
Baccatin III, ≥95% (HPLC)
Sigma-Aldrich
Gamborg′s Vitamin Solution, 1000 ×, liquid, suitable for plant cell culture
Sigma-Aldrich
Gamborg′s B-5 Basal Medium with Minimal Organics, fine powder, suitable for plant cell culture