
Zinc sulfate monohydrate

Synonym(s):
Zinc sulfate monohydrate
Empirical Formula (Hill Notation):
O4SZn · xH2O
CAS Number:
Molecular Weight:
161.45 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing