
Uridine 5′-triphosphate tris salt

Synonym(s):
UTP
Empirical Formula (Hill Notation):
C9H15N2O15P3
CAS Number:
Molecular Weight:
484.14
Products
Product Category
Brand
Formula Weight
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing