
Tryptophan Related Compound A

Synonym(s):
1,1′-Ethylidenebis-(L-tryptophan), EBT
Empirical Formula (Hill Notation):
C24H26N4O4
CAS Number:
Molecular Weight:
434.49
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2