
Triisopropylsilane

Synonym(s):
Triisopropylsilane
Linear Formula:
((CH3)2CH)3SiH
CAS Number:
Molecular Weight:
158.36
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing