Skip to Content
Merck
trans-2-Hexen-1-al natural, ≥95%, FG

trans-2-Hexen-1-al

trans-2-Hexen-1-al natural, ≥95%, FG
Linear Formula:
CH3CH2CH2CH=CHCHO
CAS Number:
Molecular Weight:
98.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Chemical Composition
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing