trans-1,3-Dichloropropene
Empirical Formula (Hill Notation):
C3H4Cl2
CAS Number:
Molecular Weight:
110.97
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1