
Tramadol hydrochloride

Synonym(s):
(±)-cis-2-(Dimethylaminomethyl)-1-(3-methoxyphenyl)cyclohexanol hydrochloride
Empirical Formula (Hill Notation):
C16H25NO2 · HCl
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3