
Tetrahydrothiophene

Synonym(s):
Tetramethylene sulfide, Thiolane, Thiophane, Tetrahydrothiophene
Empirical Formula (Hill Notation):
C4H8S
CAS Number:
Molecular Weight:
88.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing