
Sulfachlorpyridazine

Synonym(s):
Sulfachloropyridazine, 4-Amino-N-(6-chloro-3-pyridazinyl)benzenesulfonamide, N1-(6-Chloro-3-pyridazinyl)sulfanilamide
Empirical Formula (Hill Notation):
C10H9ClN4O2S
CAS Number:
Molecular Weight:
284.72
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2