
Sodium D-gluconate

Synonym(s):
D-Gluconate sodium salt, D-Gluconic acid sodium salt
Linear Formula:
HOCH2[CH(OH)]4CO2Na
CAS Number:
Molecular Weight:
218.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing