
Sodium Acetate

Synonym(s):
Sodium acetate trihydrate, Acetic acid sodium salt trihydrate
Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Products
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing